4-(2-Naphthyl)piperidine hydrochloride ≥95%

Supplier: Matrix Scientific

054817-500MG 054817-1G
101908-652EA 747.69 USD
101908-652 101908-654
4-(2-Naphthyl)piperidine hydrochloride ≥95%
4-(2-Naphthyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₈ClN
MDL Number: MFCD02179087
CAS Number: 198334-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR