N-[3-(Morpholin-4-yl)propyl]piperidine-4-carboxamide ≥95%

Supplier: Matrix Scientific

054815-500MG 054815-1G
101908-644EA 1109.23 USD
101908-644 101908-646
N-[3-(Morpholin-4-yl)propyl]piperidine-4-carboxamide ≥95%
N-[3-(Morpholin-4-yl)propyl]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₃H₂₅N₃O₂
MDL Number: MFCD08442442
CAS Number: 926255-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR