8-Methyl-1-thia-4-azaspiro[4.5]decane ≥95%

Supplier: Matrix Scientific

054813-500MG 054813-1G
101908-634EA 278.46 USD
101908-634 101908-636
8-Methyl-1-thia-4-azaspiro[4.5]decane ≥95%
8-Methyl-1-thia-4-azaspiro[4.5]decane

Hazard: Irritant

Formula: C₉H₁₇NS
MDL Number: MFCD16140310
CAS Number: 31404-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR