(1-Methyl-1,4,5,6-tetrahyrdopyrimidin-2-yl)methylamine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (1-Methyl-1,4,5,6-tetrahydropyrimidin-2-yl)methanamine dihydrochloride

054812-500MG 054812-1G
101908-630EA 747.69 USD
101908-630 101908-632
(1-Methyl-1,4,5,6-tetrahyrdopyrimidin-2-yl)methylamine dihydrochloride ≥95%
(1-Methyl-1,4,5,6-tetrahyrdopyrimidin-2-yl)methylamine dihydrochloride

Hazard: Irritant

Formula: C₆H₁₅Cl₂N₃
MW: 200.11 g/mol
MDL Number: MFCD16090014
CAS Number: 73706-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR