1-(Methylsulfonyl)piperidine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

054806-500MG 054806-1G
101908-606EA 376.92 USD
101908-606 101908-608
1-(Methylsulfonyl)piperidine-2-carboxylic acid ≥95%
1-(Methylsulfonyl)piperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₃NO₄S
MDL Number: MFCD07357409
CAS Number: 1008975-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR