4-(4-(Methylsulfonyl)piperazin-1-yl)phenol ≥95%

Supplier: Matrix Scientific

054804-500MG 054804-1G
101908-598EA 556.92 USD
101908-598 101908-600
4-(4-(Methylsulfonyl)piperazin-1-yl)phenol ≥95%
4-(4-(Methylsulfonyl)piperazin-1-yl)phenol

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃S
MDL Number: MFCD16090027
CAS Number: 67915-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR