1-(Methylsulfonyl)-3-piperidinol ≥95%

Supplier: Matrix Scientific

054803-500MG 054803-1G
101908-594EA 747.69 USD
101908-594 101908-596
1-(Methylsulfonyl)-3-piperidinol ≥95%
1-(Methylsulfonyl)-3-piperidinol

Hazard: Irritant

Formula: C₆H₁₃NO₃S
MDL Number: MFCD12185766
CAS Number: 444606-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR