N-(Methylsulfonyl)-DL-phenylalanine ≥95%

Supplier: Matrix Scientific

054802-500MG 054802-1G
101908-590EA 747.69 USD
101908-590 101908-592
N-(Methylsulfonyl)-DL-phenylalanine ≥95%
N-(Methylsulfonyl)-DL-phenylalanine

Hazard: Irritant

Formula: C₁₀H₁₃NO₄S
MDL Number: MFCD06434623
CAS Number: 29268-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR