2-(1-Methyl-1H-imidazol-2-ylthio)acetic acid ≥95%

Supplier: Matrix Scientific

054796-500MG 054796-1G
101908-566EA 710.77 USD
101908-566 101908-568
2-(1-Methyl-1H-imidazol-2-ylthio)acetic acid ≥95%
2-(1-Methyl-1H-imidazol-2-ylthio)acetic acid

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MDL Number: MFCD02257972
CAS Number: 71370-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR