2-(3-Methoxyphenoxy)ethanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(3-Methoxyphenoxy)ethylamine

054783-500MG 054783-1G
101908-510EA 153.85 USD
101908-510 101908-512
2-(3-Methoxyphenoxy)ethanamine ≥95%
2-(3-Methoxyphenoxy)ethanamine

Hazard: Irritant

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD02598966
CAS Number: 6487-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR