3-Methanesulfonylpropylammonium chloride 95
Supplier: Matrix Scientific
054780-500MG
054780-1G
101908-500EA
538.46
USD
101908-500
101908-502
3-Methanesulfonylpropylammonium chloride 95
3-Methanesulfonylpropylammonium chloride
Hazard: Irritant
Formula:
C₄H₁₂ClNO₂S MW: 173.66 g/mol |
MDL Number:
MFCD09744144 CAS Number: 26209-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...