1-(2-Hydroxyphenyl)piperazin-4-methylsulfonamide ≥95%

Supplier: Matrix Scientific

054775-500MG 054775-1G
101908-478EA 556.92 USD
101908-478 101908-480
1-(2-Hydroxyphenyl)piperazin-4-methylsulfonamide ≥95%
1-(2-Hydroxyphenyl)piperazin-4-methylsulfonamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃S
MDL Number: MFCD16090023
CAS Number: 1221792-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR