1-(3-Hydroxyphenyl)-4-ethoxycarbonylpiperazine ≥95%

Supplier: Matrix Scientific

054772-500MG 054772-1G
101908-466EA 556.92 USD
101908-466 101908-468
1-(3-Hydroxyphenyl)-4-ethoxycarbonylpiperazine ≥95%
1-(3-Hydroxyphenyl)-4-ethoxycarbonylpiperazine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD16090025
CAS Number: 67915-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR