1-(2-Hydroxyethyl)benzimidazole ≥95%

Supplier: Matrix Scientific

054768-500MG 054768-1G
101908-450EA 278.46 USD
101908-450 101908-452
1-(2-Hydroxyethyl)benzimidazole ≥95%
1-(2-Hydroxyethyl)benzimidazole

Hazard: Irritant

Formula: C₉H₁₀N₂O
MDL Number: MFCD00483391
CAS Number: 6340-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR