N-[4-(Hydroxyethoxy)phenyl]ethanamide ≥95%

Supplier: Matrix Scientific

054767-500MG 054767-1G
101908-446EA 921.54 USD
101908-446 101908-448
N-[4-(Hydroxyethoxy)phenyl]ethanamide ≥95%
N-[4-(Hydroxyethoxy)phenyl]ethanamide

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00020595
CAS Number: 50375-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR