FMOC-DL-3-thienylalanine ≥95%

Supplier: Matrix Scientific

054763-500MG 054763-1G
101908-430EA 921.54 USD
101908-430 101908-432
FMOC-DL-3-thienylalanine ≥95%
FMOC-DL-3-thienylalanine

Hazard: Irritant

Formula: C₂₂H₁₉NO₄S
MDL Number: MFCD06656862
CAS Number: 678991-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR