Fmoc-DL-4-tert-butyl-Phe ≥95%
Supplier: Matrix Scientific
054762-500MG
054762-1G
101908-426EA
921.54
USD
101908-426
101908-428
Fmoc-DL-4-tert-butyl-Phe ≥95%
Fmoc-DL-4-tert-butyl-Phe
Hazard: Irritant
Formula:
C₂₈H₂₉NO₄ |
MDL Number:
MFCD16622825 CAS Number: 1236257-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...