FMOC-DL-styrylalanine ≥95%

Supplier: Matrix Scientific

054761-500MG 054761-1G
101908-422EA 921.54 USD
101908-422 101908-424
FMOC-DL-styrylalanine ≥95%
FMOC-DL-styrylalanine

Hazard: Irritant

Formula: C₂₆H₂₃NO₄
MW: 413.48 g/mol
MDL Number: MFCD02094011
CAS Number: 1236297-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR