Fmoc-DL-2-pyridylalanine ≥95%

Supplier: Matrix Scientific

054760-500MG 054760-1G
101908-418EA 921.54 USD
101908-418 101908-420
Fmoc-DL-2-pyridylalanine ≥95%
Fmoc-DL-2-pyridylalanine

Hazard: Irritant

Formula: C₂₃H₂₀N₂O₄
MDL Number: MFCD02094443
CAS Number: 1236267-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR