Fmoc-DL-4-pyridylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(pyridin-4-yl)propanoic acid, Fmoc-DL-3-(4-Pyridyl)-Ala-OH

054759-500MG 054759-1G
101908-414EA 921.54 USD
101908-414 101908-416
Fmoc-DL-4-pyridylalanine ≥95%
Fmoc-DL-4-pyridylalanine

Hazard: Irritant

Formula: C₂₃H₂₀N₂O₄
MW: 388.43 g/mol
MDL Number: MFCD03701331
CAS Number: 746672-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR