FMOC-DL-2-furylalanine ≥95%

Supplier: Matrix Scientific

054758-500MG 054758-1G
101908-410EA 921.54 USD
101908-410 101908-412
FMOC-DL-2-furylalanine ≥95%
FMOC-DL-2-furylalanine

Hazard: Irritant

Formula: C₂₂H₁₉NO₅
MW: 377.4 g/mol
MDL Number: MFCD02094001
CAS Number: 1236257-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR