Fmoc-DL-3,3-diphenylalanine ≥95%

Supplier: Matrix Scientific

054757-500MG 054757-1G
101908-406EA 921.54 USD
101908-406 101908-408
Fmoc-DL-3,3-diphenylalanine ≥95%
Fmoc-DL-3,3-diphenylalanine

Hazard: Irritant

Formula: C₃₀H₂₅NO₄
MW: 463.54 g/mol
MDL Number: MFCD02093614
CAS Number: 839719-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR