Fmoc-β-(3-benzothienyl)-Ala-OH ≥95%

Supplier: Matrix Scientific

Synonyms: Fmoc-3-Ala(3-benzothienyl)-OH, Fmoc-β-(3-benzothienyl)-Ala-OH

054756-500MG 054756-1G
101908-402EA 921.54 USD
101908-402 101908-404
Fmoc-β-(3-benzothienyl)-Ala-OH ≥95%
Fmoc-beta-(3-benzothienyl)-Ala-OH

Hazard: Irritant

Formula: C₂₆H₂₁NO₄S
MW: 443.517 g/mol
MDL Number: MFCD00672562
CAS Number: 177966-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR