N-(Ethylsulfonyl)-4-aminobutyric acid ≥95%

Supplier: Matrix Scientific

054750-500MG 054750-1G
101908-382EA 747.69 USD
101908-382 101908-384
N-(Ethylsulfonyl)-4-aminobutyric acid ≥95%
N-(Ethylsulfonyl)-4-aminobutyric acid

Hazard: Irritant

Formula: C₆H₁₃NO₄S
MDL Number: MFCD08443567
CAS Number: 926247-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR