Ethyl (1R,4R)-4-hydroxycyclohexylcarbamate ≥95%

Supplier: Matrix Scientific

054749-500MG 054749-1G
101908-378EA 1855.38 USD
101908-378 101908-380
Ethyl (1R,4R)-4-hydroxycyclohexylcarbamate ≥95%
Ethyl (1R,4R)-4-hydroxycyclohexylcarbamate

Hazard: Irritant

Formula: C₉H₁₇NO₃
MDL Number: MFCD09836369
CAS Number: 71118-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR