1-Ethyl-3-((1R,4R)-4-hydroxycyclohexyl)urea ≥95%

Supplier: Matrix Scientific

054748-500MG 054748-1G
101908-374EA 1855.38 USD
101908-374 101908-376
1-Ethyl-3-((1R,4R)-4-hydroxycyclohexyl)urea ≥95%
1-Ethyl-3-((1R,4R)-4-hydroxycyclohexyl)urea

Hazard: Irritant

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD09836380
CAS Number: 1241675-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR