N-Ethyl-4-hydroxypiperidine-1-carboxamide ≥95%

Supplier: Matrix Scientific

054746-500MG 054746-1G
101908-368EA 709.23 USD
101908-368 101908-370
N-Ethyl-4-hydroxypiperidine-1-carboxamide ≥95%
N-Ethyl-4-hydroxypiperidine-1-carboxamide

Hazard: Irritant

Formula: C₈H₁₆N₂O₂
MDL Number: MFCD12185727
CAS Number: 1156104-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR