1-Ethyl-3-(5-hydroxypentyl)urea ≥95%

Supplier: Matrix Scientific

054745-500MG 054745-1G
101908-364EA 747.69 USD
101908-364 101908-366
1-Ethyl-3-(5-hydroxypentyl)urea ≥95%
1-Ethyl-3-(5-hydroxypentyl)urea

Hazard: Irritant

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD16090038
CAS Number: 939901-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR