N-Ethyl-2-(hydroxymethyl)piperidine-1-carboxamide ≥95%

Supplier: Matrix Scientific

054742-500MG 054742-1G
101908-352EA 747.69 USD
101908-352 101908-354
N-Ethyl-2-(hydroxymethyl)piperidine-1-carboxamide ≥95%
N-Ethyl-2-(hydroxymethyl)piperidine-1-carboxamide

Hazard: Irritant

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD12141719
CAS Number: 939909-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR