N-Ethyl-2-(2-hydroxyethyl)piperidine-1-carboxamide ≥95%

Supplier: Matrix Scientific

054741-500MG 054741-1G
101908-348EA 747.69 USD
101908-348 101908-350
N-Ethyl-2-(2-hydroxyethyl)piperidine-1-carboxamide ≥95%
N-Ethyl-2-(2-hydroxyethyl)piperidine-1-carboxamide

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD12813110
CAS Number: 1183246-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR