Ethyl-5-(cyanomethyl)-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

054738-500MG 054738-1G
101908-334EA 384.62 USD
101908-334 101908-336
Ethyl-5-(cyanomethyl)-2-pyridinecarboxylate ≥95%
Ethyl-5-(cyanomethyl)-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
MDL Number: MFCD16140285
CAS Number: 913839-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR