Ethyl-2-chloro-4-(trifluoromethyl)nicotinate ≥95%

Supplier: Matrix Scientific

054737-500MG 054737-1G
101908-330EA 710.77 USD
101908-330 101908-332
Ethyl-2-chloro-4-(trifluoromethyl)nicotinate ≥95%
Ethyl-2-chloro-4-(trifluoromethyl)nicotinate

Hazard: Irritant

Formula: C₉H₇ClF₃NO₂
MDL Number: MFCD16140284
CAS Number: 1221792-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR