3-(3-[(Ethylamino)carbonyl]aminophenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

054735-500MG 054735-1G
101908-322EA 747.69 USD
101908-322 101908-324
3-(3-[(Ethylamino)carbonyl]aminophenyl)propanoic acid ≥95%
3-(3-[(Ethylamino)carbonyl]aminophenyl)propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD16090019
CAS Number: 1221792-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR