3-[(Ethylamino)carbonyl]aminobenzoic acid ≥95%

Supplier: Matrix Scientific

054734-500MG 054734-1G
101908-318EA 921.54 USD
101908-318 101908-320
3-[(Ethylamino)carbonyl]aminobenzoic acid ≥95%
3-[(Ethylamino)carbonyl]aminobenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD08310637
CAS Number: 23754-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR