1-(Ethoxycarbonyl)piperidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

054732-500MG 054732-1G
101908-310EA 556.92 USD
101908-310 101908-312
1-(Ethoxycarbonyl)piperidine-3-carboxylic acid ≥95%
1-(Ethoxycarbonyl)piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄
MDL Number: MFCD09816343
CAS Number: 944280-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR