N-(Ethoxycarbonyl)-DL-alanine ≥95%
Supplier: Matrix Scientific
054730-500MG
054730-1G
101908-302EA
747.69
USD
101908-302
101908-304
N-(Ethoxycarbonyl)-DL-alanine ≥95%
N-(Ethoxycarbonyl)-DL-alanine
Hazard: Irritant
Formula:
C₆H₁₁NO₄ MW: 161.16 g/mol |
MDL Number:
MFCD09816240 CAS Number: 6318-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...