N-(Ethoxycarbonyl)-4-aminobutyric acid ≥95%

Supplier: Matrix Scientific

054729-500MG 054729-1G
101908-298EA 747.69 USD
101908-298 101908-300
N-(Ethoxycarbonyl)-4-aminobutyric acid ≥95%
N-(Ethoxycarbonyl)-4-aminobutyric acid

Hazard: Irritant

Formula: C₇H₁₃NO₄
Melting Pt: 43…45 °C
MDL Number: MFCD00036915
CAS Number: 4143-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR