2-{4-[(Ethoxycarbonyl)amino]phenyl}acetic acid ≥95%

Supplier: Matrix Scientific

054728-500MG 054728-1G
101908-294EA 747.69 USD
101908-294 101908-296
2-{4-[(Ethoxycarbonyl)amino]phenyl}acetic acid ≥95%
2-{4-[(Ethoxycarbonyl)amino]phenyl}acetic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD09807246
CAS Number: 91134-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR