Dimethyl(4-pyrrolidin-2-ylphenyl)aminemonoacetate ≥95%

Supplier: Matrix Scientific

Synonyms: N,N-dimethyl-4-(pyrrolidin-2-yl)aniline

054726-500MG 054726-1G
101908-286EA 747.69 USD
101908-286 101908-288
Dimethyl(4-pyrrolidin-2-ylphenyl)aminemonoacetate ≥95%
Dimethyl(4-pyrrolidin-2-ylphenyl)aminemonoacetate

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₂
MDL Number: MFCD06660377
CAS Number: 298690-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR