4-(Dimethylcarbamoyloxy)phenylacetic acid ≥95%

Supplier: Matrix Scientific

054724-500MG 054724-1G
101908-278EA 375.38 USD
101908-278 101908-280
4-(Dimethylcarbamoyloxy)phenylacetic acid ≥95%
4-(Dimethylcarbamoyloxy)phenylacetic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD03411192
CAS Number: 1221791-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR