2-((Dimethylcarbamoyl)methoxy)benzoic acid ≥95%

Supplier: Matrix Scientific

054723-500MG 054723-1G
101908-274EA 278.46 USD
101908-274 101908-276
2-((Dimethylcarbamoyl)methoxy)benzoic acid ≥95%
2-((Dimethylcarbamoyl)methoxy)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD07314040
CAS Number: 847783-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR