(N,N-Dimethyl)(3-aminopropyl)sulfonamide hydrochloride ≥95%

Supplier: Matrix Scientific

054722-500MG 054722-1G
101908-270EA 921.54 USD
101908-270 101908-272
(N,N-Dimethyl)(3-aminopropyl)sulfonamide hydrochloride ≥95%
(N,N-Dimethyl)(3-aminopropyl)sulfonamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₅ClN₂O₂S
MDL Number: MFCD16090034
CAS Number: 91893-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR