4-(3-(Dimethylamino)-3-oxopropyl)benzoic acid ≥95%

Supplier: Matrix Scientific

054721-500MG 054721-1G
101908-266EA 556.92 USD
101908-266 101908-268
4-(3-(Dimethylamino)-3-oxopropyl)benzoic acid ≥95%
4-(3-(Dimethylamino)-3-oxopropyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD09836504
CAS Number: 937642-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR