3-(3-(Dimethylamino)-3-oxopropyl)benzoic acid ≥95%

Supplier: Matrix Scientific

054720-500MG 054720-1G
101908-262EA 556.92 USD
101908-262 101908-264
3-(3-(Dimethylamino)-3-oxopropyl)benzoic acid ≥95%
3-(3-(Dimethylamino)-3-oxopropyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD16090033
CAS Number: 940099-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR