2-(4-{[(Dimethylamino)carbonyl]amino}phenyl)acetic acid ≥95%

Supplier: Matrix Scientific

054718-500MG 054718-1G
101908-254EA 747.69 USD
101908-254 101908-256
2-(4-{[(Dimethylamino)carbonyl]amino}phenyl)acetic acid ≥95%
2-(4-{[(Dimethylamino)carbonyl]amino}phenyl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃
MDL Number: MFCD14596096
CAS Number: 1221792-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR