1-(2,5-Dichloro-3-pyridinyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

054710-500MG 054710-1G
101908-228EA 747.69 USD
101908-228 101908-230
1-(2,5-Dichloro-3-pyridinyl)-1-ethanone ≥95%
1-(2,5-Dichloro-3-pyridinyl)-1-ethanone

Hazard: Irritant

Formula: C₇H₅Cl₂NO
MDL Number: MFCD16140278
CAS Number: 594815-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR