6-[(4-Chlorophenyl)sulfanyl]-3-pyridinylamine ≥95%

Supplier: Matrix Scientific

054704-500MG 054704-1G
101908-202EA 556.92 USD
101908-202 101908-204
6-[(4-Chlorophenyl)sulfanyl]-3-pyridinylamine ≥95%
6-[(4-Chlorophenyl)sulfanyl]-3-pyridinylamine

Hazard: Irritant

Formula: C₁₁H₉ClN₂S
MW: 236.72 g/mol
MDL Number: MFCD00052804
CAS Number: 25935-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR