5-(Chloromethyl)-1-phenyl-1H-1,2,4-triazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

054702-500MG 054702-1G
101908-196EA 660 USD
101908-196 101908-198
5-(Chloromethyl)-1-phenyl-1H-1,2,4-triazole-3-carboxylic acid ≥95%
5-(Chloromethyl)-1-phenyl-1H-1,2,4-triazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈ClN₃O₂
MDL Number: MFCD16090020
CAS Number: 1221792-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR