6-Chloro-N-methoxy-N-methylnicotinamide ≥95%

Supplier: Matrix Scientific

054700-500MG 054700-1G 054700-5G
101908-186EA 278.46 USD
101908-186 101908-188 101908-190
6-Chloro-N-methoxy-N-methylnicotinamide ≥95%
6-Chloro-N-methoxy-N-methylnicotinamide

Hazard: Irritant

Formula: C₈H₉ClN₂O₂
MDL Number: MFCD09991647
CAS Number: 149281-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR