N-Chloroacetyl-4-aminophenylpropionic acid ≥95%

Supplier: Matrix Scientific

054697-500MG 054697-1G
101908-174EA 556.92 USD
101908-174 101908-176
N-Chloroacetyl-4-aminophenylpropionic acid ≥95%
N-Chloroacetyl-4-aminophenylpropionic acid

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃
MDL Number: MFCD16140272
CAS Number: 500336-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR